EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Structural influence of ordered mesoporous carbon supports for the hydrogenation of carbon monoxide to alcohols.
PMID 24245283
Abstract
A series of ordered mesoporous carbon materials (OMCs) possessing well-ordered nanoporosity with different mesopore structures were synthesized by the template-synthesis route. Two different pore strucutes (2-dimensional hexagonal and 3-dimensional cubic structures) and two different framework-configurations (rod-type and hollow-type carbon frameworks) are prepared by using the two different silica templates and synthetic conditions. The ordered mesoporous carbon supported promoted-rhodium catalysts were preparted by an incipient wetness method. The promoted Rh-OMC catalysts are tested by a fixed bed reactor for the catalytic conversion of syngas-to-alcohols. The characteristics of the promoted Rh-OMCs catalysts were scrutinized through a series of different techniques, including transmission electron microscopy (TEM), powder X-ray diffraction (XRD), and N2 sorption analysis, and the catalytic performance was tested in a fixed-bed reactor. It was found that the promoted Rh-OMC catalysts exhibited the different catalytic activity and selectivity of alcohols, which could be attributed to the size of metal nanoparticles being confined by the different mesostructure of OMCs.