EMAIL THIS PAGE TO A FRIEND
Psychiatria Danubina
Treatment of severe sleep disorder related to alcohol-dependence with high-dose agomelatine - a case report.
PMID 24247055
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1362
Agomelatine, ≥98% (HPLC)
C15H17NO2