EMAIL THIS PAGE TO A FRIEND
Molecular Carcinogenesis
Benzo[a]pyrene-7,8-diol-9,10-epoxide inhibits gap junction intercellular communication via phosphorylation of tumor progression locus 2 in WB-F344 rat liver epithelial cells.
PMID 24249418