EMAIL THIS PAGE TO A FRIEND
Experimental mechanics
Following the Behavior of the Flagellar Rotary Motor Near Zero Load.
PMID 24249872
Abstract
At room temperature at stall, the flagellar motor of the bacterium