EMAIL THIS PAGE TO A FRIEND
Organic letters
Iron porphyrin-catalyzed three-component reaction of ethyl diazoacetate with aliphatic amines and β,γ-unsaturated α-keto esters.
PMID 24251920
Abstract
An iron porphyrin-catalyzed three-component reaction of ethyl diazoacetate with aliphatic amines and β,γ-unsaturated α-keto esters is reported. The use of iron porphyrin catalyst allows aliphatic amines to be used as the substrate without encountering catalyst poisoning issue and a series of β-hydroxy-α-amino esters are produced in high yields with excellent regioselectivities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E22201
Ethyl diazoacetate, contains ≥13 wt. % dichloromethane
C4H6N2O2