EMAIL THIS PAGE TO A FRIEND
Zhonghua jie he he hu xi za zhi = Zhonghua jiehe he huxi zazhi = Chinese journal of tuberculosis and respiratory diseases
[The analysis of clinical data about the treatment of linezolid against tuberculosis].
PMID 24252739
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0014
Linezolid, ≥98% (HPLC)
C16H20FN3O4