EMAIL THIS PAGE TO A FRIEND
Mucosal immunology
Gal-lectin-dependent contact activates the inflammasome by invasive Entamoeba histolytica.
PMID 24253103
Abstract
Entamoeba histolytica (Eh) is an extracellular protozoan parasite of the human colon, which occasionally breaches the intestinal barrier. Eradicating ameba that invades is essential for host survival. A defining but uncharacterized feature of amebic invasion is direct contact between ameba and host cells. This event corresponds with a massive pro-inflammatory response. To date, pathogen recognition receptors (PRRs) that are activated by contact with viable Eh are unknown. Here we show that the innate immune system responds in a qualitatively different way to contact with viable Eh vs. soluble ligands produced by viable or dead ameba. This unique Eh Gal-lectin contact-dependent response in macrophages was mediated by activation of the inflammasome. Soluble native Gal-lectin did not induce inflammasome activation, but was sufficient for transcriptional priming of the inflammasome and non-inflammasome-dependent pro-inflammatory cytokine release. We conclude the inflammasome is a pathogenicity sensor for invasive Eh and identify for the first time a PRR that specifically responds to contact with intact parasites in a manner that accords with scale immune response to parasite invasion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA003056
Anti-CASP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS3459
Anti-Caspase-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3463
Anti-Caspase-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
G0750
D-(+)-Galactose, ≥99%
C6H12O6
G0625
D-(+)-Galactose, ≥98%
C6H12O6
G6404
D-(+)-Galactose, BioXtra, pH 5.0-7.0 (20 °C, 1 M in H2O), ≥99%
C6H12O6
G5388
D-(+)-Galactose, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture
C6H12O6
48260
D-(+)-Galactose, for microbiology, ≥99.0%
C6H12O6
15522
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
C6H12O6
1287700 Galactose, United States Pharmacopeia (USP) Reference Standard
C6H12O6
PHR1206
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6
G0050000
Galactose, European Pharmacopoeia (EP) Reference Standard
C6H12O6
ARK2176
Galactose, plant-derived, PharmaGrade, USP/NF, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C6H12O6
WH0000834M1
Monoclonal Anti-CASP1 antibody produced in mouse, clone 3D2, purified immunoglobulin, buffered aqueous solution