EMAIL THIS PAGE TO A FRIEND
European review for medical and pharmacological sciences
Computer-assisted implant surgery and immediate loading in edentulous ridges with dental fresh extraction sockets. Two years results of a prospective case series study.
PMID 24254569
Abstract
The two-stage surgical approach for implant placement first documented in 1977 by Brånemark, represents today the most used protocol for placing implants. Aim of this prospective case series study was to compare the clinical and radiological performance of 12 edentulous jaws treated with of a modified prosthetic and surgical protocol for 3D software planning, guided surgery, immediate loading of implants inserted in edentulous jaws and extraction sockets and restored with Cad-Cam Zirconia and titanium full arch frameworks. This work was designed as a prospective case series study. Twelve patients have been consecutively rehabilitated with an immediately loaded implant supported fixed full prosthesis. A total of 72 implants, Nobel Replace Tapered Groovy; Nobel Biocare AB, Goteborg, Sweden) 26 of which were inserted in fresh extraction sockets, were inserted. Outcome measures were implants survival, radiographic marginal bone-levels and bone remodeling, soft tissue parameters and complications. All patients reached 24 months follow-up, and no patients dropped out from the study. The cumulative survival rate was 100%; after 24 months mean marginal bone remodelling value was: 1.35 ± 0.25, mean PPD value was 2.75 ± 0.40 mm and mean BOP value was 3.8% ± 1.8%. Only minor prosthetic complications were recorded. These data seem to validate this surgical and prosthetic protocol with valid results when applied in selected cases.