EMAIL THIS PAGE TO A FRIEND
Clinical science (London, England : 1979)
Suppression of abdominal aortic aneurysm formation by inhibition of prolyl hydroxylase domain protein through attenuation of inflammation and extracellular matrix disruption.
PMID 24256203
Abstract
In the present study we sought to determine the effect of CoCl2, an inhibitor of PHD (prolyl hydroxylase domain protein), on the development of AAA (abdominal aortic aneurysm). AAA was induced in C57BL/6 mice by periaortic application of CaCl2 (AAA group). NaCl (0.9%)-treated mice were used as a sham control (SHAM group). Mice were treated with 0.05% CoCl2 in the drinking water (AAA/CoCl2 group). At 1 and 6 weeks after the operation, aortic tissue was excised for further examination. After 6 weeks of CaCl2 treatment, aortic diameter and macrophage infiltration into the aortic adventitia were increased in the AAA group compared with the SHAM group. Treatment with CoCl2 reduced the aneurysmal size and macrophage infiltration compared with the AAA group. Aortic expression of inflammatory cytokines and MCP-1 (monocyte chemoattractant protein-1) and the activities of MMP-9 (matrix metalloproteinase-9) and MMP-2 were enhanced in the AAA group and attenuated in the AAA/CoCl2 group. Expression of cytokines and the activities of MMPs were already increased after 1 week of CaCl2 treatment, but were suppressed by CoCl2 treatment in association with reduced NF-κB (nuclear factor κB) phosphorylation. Treatment with CoCl2 in mice prevented the development of CaCl2-induced AAA in association with reduced inflammation and ECM (extracellular matrix) disruption. The results of the present study suggest that PHD plays a critical role in the development of AAA and that there is a therapeutic potential for PHD inhibitors in the prevention of AAA development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
223506
Calcium chloride dihydrate, ACS reagent, ≥99%
CaCl2 · 2H2O
C3881
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
CaCl2 · 2H2O
31307
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
CaCl2 · 2H2O
12022
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
CaCl2 · 2H2O
21102
Calcium chloride dihydrate, tested according to Ph Eur
CaCl2 · 2H2O
21098
Calcium chloride dihydrate, BioUltra, ≥99.5% (KT)
CaCl2 · 2H2O
GF76259593
Cobalt, foil, 100x100mm, thickness 2.0mm, as rolled, 99.9%
Co
GF48813723
Cobalt, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Co
GF13761918
Cobalt, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Co
GF95441283
Cobalt, foil, 50x50mm, thickness 0.5mm, as rolled, 99.99+%
Co
GF74432392
Cobalt, wire reel, 0.1m, diameter 0.05mm, hard, 99.9%
Co
GF20239017
Cobalt, wire reel, 1m, diameter 0.5mm, hard, 99.99+%
Co
GF41834011
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.9%
Co
GF85563535
Cobalt, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Co
GF55186050
Cobalt, wire reel, 0.2m, diameter 1.0mm, hard, 99.99+%
Co
GF12274253
Cobalt, wire reel, 0.1m, diameter 2.0mm, hard, 99.99+%
Co
GF70398502
Cobalt, wire reel, 0.2m, diameter 0.25mm, hard, 99.99+%
Co
GF29762646
Cobalt, foil, 4mm disks, thickness 0.025mm, 99.9%
Co
GF91564904
Cobalt, foil, 250x250mm, thickness 0.50mm, annealed, 99.9%
Co
GF90510094
Cobalt, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%
Co
GF15401135
Cobalt, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Co
GF63298198
Cobalt, wire reel, 0.05m, diameter 2.0mm, hard, 99.99+%
Co
GF54906385
Cobalt, wire reel, 0.025m, diameter 2.0mm, hard, 99.99+%
Co
GF15124081
Cobalt, wire reel, 0.1m, diameter 0.5mm, hard, 99.99+%
Co
GF60120997
Cobalt, wire reel, 0.05m, diameter 0.5mm, hard, 99.99+%
Co
GF91382069
Cobalt, wire reel, 0.1m, diameter 0.1mm, hard, 99.99+%
Co
GF94594450
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
Co
GF41541476
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Co
GF45372315
Cobalt, foil, 4mm disks, thickness 0.003mm, 99.9%
Co
GF08560289
Cobalt, foil, 4mm disks, thickness 0.004mm, 99.9%
Co