EMAIL THIS PAGE TO A FRIEND
JAMA dermatology
Isotretinoin and pregnancy prevention: do we need to take a long, hard look at ourselves?
PMID 24258339
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1353500
Isotretinoin, United States Pharmacopeia (USP) Reference Standard
C20H28O2