EMAIL THIS PAGE TO A FRIEND
JAMA dermatology
Women's experiences with isotretinoin risk reduction counseling.
PMID 24258663
Abstract
Isotretinoin, an effective anti-acne therapy, is a known teratogen that is strictly regulated through the iPLEDGE program. However, since this program has not significantly reduced rates of pregnancies exposed to isotretinoin, new strategies for reducing rates of isotretinoin-exposed pregnancies are needed. To explore women's experiences with counseling about isotretinoin risk reduction. Structured interviews were conducted between January and September 2012. Two independent coders performed content analysis using a grounded theory approach. The study participants were 16 women who had used isotretinoin to treat severe skin disease and who were recruited from a single urban community via flyers displayed on college campuses, at dermatology clinics, and at student health facilities. Perceptions of isotretinoin-associated risks and understanding of ways teratogenic risks can be avoided. Participants clearly understood that isotretinoin is teratogenic but had less understanding of contraceptive methods that effectively prevent pregnancy. Most described the counseling they received as anxiety provoking. Few were counseled about highly effective reversible contraceptives such as the subdermal implant or intrauterine contraception; most counseling focused on oral contraceptives. Women cited multiple influences on their contraceptive choices, including friends, family, physicians, the internet, and other media; however, some expressed concerns about the accuracy of these sources of information. For many, iPLEDGE was their first introduction to contraception. When presented with evidence-based information on the relative effectiveness of available contraceptives, participants expressed surprise that this was not part of the iPLEDGE materials. Since few clinicians provide women information on highly effective (ie, intrauterine or subdermal) contraceptives, the iPLEDGE program increases anxiety about isotretinoin more than it helps women feel protected from the teratogenic risks of isotretinoin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1353500
Isotretinoin, United States Pharmacopeia (USP) Reference Standard
C20H28O2