EMAIL THIS PAGE TO A FRIEND
Food chemistry
Recovery and quantitative detection of thiabendazole on apples using a surface swab capture method followed by surface-enhanced Raman spectroscopy.
PMID 24262524
Abstract
We developed a rapid and simple method which combines a surface swab capture method and surface-enhanced Raman spectroscopy for recovery and quantitative detection of thiabendazole on apple surfaces. The whole apple surface was swabbed and the swab was vortexed in methanol releasing the pesticide. Silver dendrites were then added to bind the pesticide and used for enhancing the Raman signals. The recovery of the surface swab method was calculated to be 59.4-76.6% for intentionally contaminated apples at different levels (0.1, 0.3, 3, and 5 ppm, μg/g per weight). After considering the releasing factor (66.6%) from the swab, the final accuracy of the swab-SERS method was calculated to be between 89.2% and 115.4%. This swab-SERS method is simple, sensitive, rapid (∼10 min), and quantitative enough for QA/QC in plant procedure. This can be extended to detect other pesticides on raw agricultural produce like pears, carrots, and melons etc.