EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Selective C-H fluorination of pyridines and diazines inspired by a classic amination reaction.
PMID 24264986
Abstract
Fluorinated heterocycles are prevalent in pharmaceuticals, agrochemicals, and materials. However, reactions that incorporate fluorine into heteroarenes are limited in scope and can be hazardous. We present a broadly applicable and safe method for the site-selective fluorination of a single carbon-hydrogen bond in pyridines and diazines using commercially available silver(II) fluoride. The reactions occur at ambient temperature within 1 hour with exclusive selectivity for fluorination adjacent to nitrogen. The mild conditions allow access to fluorinated derivatives of medicinally important compounds, as well as a range of 2-substituted pyridines prepared by subsequent nucleophilic displacement of fluoride. Mechanistic studies demonstrate that the pathway of a classic pyridine amination can be adapted for selective fluorination of a broad range of nitrogen heterocycles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295396
Hydrogen, ≥99.99%
H2