EMAIL THIS PAGE TO A FRIEND
The Cochrane database of systematic reviews
Thyrotropin-releasing hormone added to corticosteroids for women at risk of preterm birth for preventing neonatal respiratory disease.
PMID 24265169
Abstract
Thyrotropin-releasing hormones (TRH) added to prenatal corticosteroids has been suggested as a way to further reduce breathing problems and neonatal lung disease in infants born preterm. To assess the effects of giving prenatal TRH in addition to corticosteroids to women at risk of preterm birth for the prevention of neonatal respiratory disease. We searched the Cochrane Pregnancy and Childbirth Group's Trials Register (30 June 2013) and reference lists of retrieved studies. We also contacted trial authors. Randomised controlled trials in women at sufficient risk of preterm birth to warrant the use of prenatal corticosteroids to promote lung maturity. TRH and corticosteroids were compared with corticosteroids, with or without placebo. All assessments of trial eligibility, risk of bias and data extractions were independently carried out by at least two review authors. Over 4600 women were recruited into the 15 trials included in the review, however two trials did not contribute any outcome data to the review. The trials had a moderate risk of bias. Overall, prenatal TRH, in addition to corticosteroids, did not reduce the risk of death prior to hospital discharge (risk ratio (RR) 1.05, 95% confidence interval (CI) 0.86 to 1.27, six trials, 3694 infants), neonatal respiratory distress syndrome (average RR 1.05, 95% CI 0.91 to 1.22, nine trials, 3833 infants), or chronic lung disease (RR 1.01, 95% CI 0.85 to 1.19, five trials, 2511 infants), and did not improve any of the secondary fetal, neonatal or childhood outcomes assessed by intention-to-treat analyses.Indeed, the data showed prenatal TRH to have adverse effects for women and their infants. All side effects reported (nausea, vomiting, light headedness, urgency of micturition, facial flushing) were significantly more likely to occur in women receiving TRH. In the infants, prenatal TRH increased the risk of needing respiratory support (RR 1.16, 95% CI 1.03 to 1.29, three trials, 1969 infants), and of having a low Apgar score at five minutes (RR 1.48, 95% CI 1.14 to 1.92, three trials, 1969 infants). Only three trials provided data on childhood follow-up, and while one trial suggested poorer outcomes for infants who were exposed to prenatal TRH, the other two trials, that assessed infants using an established developmental instrument, showed no clear differences between groups in follow-up outcomes.Sensitivity analyses by trial quality, or subgroups with differing times from entry to birth, or different dose regimens of TRH, did not change these findings. Prenatal TRH in addition to corticosteroids, given to women at risk of preterm birth, does not improve infant outcomes and can cause maternal side effects.