EMAIL THIS PAGE TO A FRIEND
Organic letters
Total synthesis of human galanin-like peptide through an aspartic acid ligation.
PMID 24266801
Abstract
Human galanin-like peptide (hGALP) is a newly discovered hypothalamic peptide that plays important roles in the regulation of food intake and energy balance. Here, we demonstrate that the aspartic acid ligation can be employed to achieve an efficient synthesis of hGALP. The total synthesis of hGALP enhances our ability to study its biology and facilitates the development of more stable analogues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11260
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
C8H12MgN2O8 · 2H2O