EMAIL THIS PAGE TO A FRIEND
Gastroenterology
American Gastroenterological Association Institute guideline on the use of thiopurines, methotrexate, and anti-TNF-α biologic drugs for the induction and maintenance of remission in inflammatory Crohn's disease.
PMID 24267474
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852678
6-Mercaptopurine monohydrate, 98%
C5H4N4S · H2O