EMAIL THIS PAGE TO A FRIEND
Cell transplantation
Propagation of human corneal endothelial cells: a novel dual media approach.
PMID 24268186
Abstract
Corneal endothelium-associated corneal blindness is the most common indication for corneal transplantation. Restorative corneal transplant surgery is the only option to reverse the blindness, but a global shortage of donor material remains an issue. There are immense clinical interests in the development of alternative treatment strategies to alleviate current reliance on donor materials. For such endeavors, ex vivo propagation of human corneal endothelial cells (hCECs) is required, but current methodology lacks consistency, with expanded hCECs losing cellular morphology to a mesenchymal-like transformation. In this study, we describe a novel dual media culture approach for the in vitro expansion of primary hCECs. Initial characterization included analysis of growth dynamics of hCECs grown in either proliferative (M4) or maintenance (M5) medium. Subsequent comparisons were performed on isolated hCECs cultured in M4 alone against cells expanded using the dual media approach. Further characterizations were performed using immunocytochemistry, quantitative real-time PCR, and gene expression microarray. At the third passage, results showed that hCECs propagated using the dual media approach were homogeneous in appearance, retained their unique polygonal cellular morphology, and expressed higher levels of corneal endothelium-associated markers in comparison to hCECs cultured in M4 alone, which were heterogeneous and fibroblastic in appearance. Finally, for hCECs cultured using the dual media approach, global gene expression and pathway analysis between confluent hCECs before and after 7-day exposure to M5 exhibited differential gene expression associated predominately with cell proliferation and wound healing. These findings showed that the propagation of primary hCECs using the novel dual media approach presented in this study is a consistent method to obtain bona fide hCECs. This, in turn, will elicit greater confidence in facilitating downstream development of alternative corneal endothelium replacement using tissue-engineered graft materials or cell injection therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9251
Adenosine, ≥99%
C10H13N5O4
A4036
Adenosine, suitable for cell culture, BioReagent
C10H13N5O4
01890
Adenosine
C10H13N5O4
A0230200
Adenosine, European Pharmacopoeia (EP) Reference Standard
C10H13N5O4
PHR1138
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
C10H13N5O4
Y0000005 Amphotericin B, European Pharmacopoeia (EP) Reference Standard
C47H73NO17
A1300000
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
C6H8O6
1043003
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
C6H8O6
PHR1008
Ascorbic Acid
C6H8O6
02672
Calcium atomic spectroscopy standard concentrate 1.00 g Ca, 1.00 g/L, for 1L standard solution, analytical standard
CaCl2
21049
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
CaCl2
02487
Chloroform, analytical standard
CHCl3
PHR1552
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
CHCl3
57803
L-Ascorbic acid, certified reference material, TraceCERT®
C6H8O6
95212
L-Ascorbic acid, tested according to Ph.Eur.
C6H8O6
PHR1316
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
CHCl3
209643
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Se
204307
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Se
229865
Selenium, powder, −100 mesh, 99.99% trace metals basis
Se
209651
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Se
HT-SE101
Selenium, pellets, < 5mm, ≥99.999%
Se
GF66395212
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Se
85886
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
C21H39N7O12 · 1.5H2SO4
S1400000
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
C21H39N7O12 · 1.5H2O4S
S6501
Streptomycin sulfate salt, powder
C21H39N7O12 · 1.5H2O4S
S1277
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
C21H39N7O12 · 1.5H2O4S
93595
Trypan Blue solution, 0.4%, for microscopy
C34H24N6Na4O14S4