Nucleic acids research
DNA transposition by protein transduction of the piggyBac transposase from lentiviral Gag precursors.
PMID 24270790
DNA transposon-based vectors have emerged as gene vehicles with a wide biomedical and therapeutic potential. So far, genomic insertion of such vectors has relied on the co-delivery of genetic material encoding the gene-inserting transposase protein, raising concerns related to persistent expression, insertional mutagenesis and cytotoxicity. This report describes potent DNA transposition achieved by direct delivery of transposase protein. By adapting integrase-deficient lentiviral particles (LPs) as carriers of the hyperactive piggyBac transposase protein (hyPBase), we demonstrate rates of DNA transposition that are comparable with the efficiency of a conventional plasmid-based strategy. Embedded in the Gag polypeptide, hyPBase is robustly incorporated into LPs and liberated from the viral proteins by the viral protease during particle maturation. We demonstrate lentiviral co-delivery of the transposase protein and vector RNA carrying the transposon sequence, allowing robust DNA transposition in a variety of cell types. Importantly, this novel delivery method facilitates a balanced cellular uptake of hyPBase, as shown by confocal microscopy, and allows high-efficiency production of clones harboring a single transposon insertion. Our findings establish engineered LPs as a new tool for transposase delivery. We believe that protein transduction methods will increase applicability and safety of DNA transposon-based vector technologies.