EMAIL THIS PAGE TO A FRIEND
International journal of clinical oncology
Expression and prognostic value of Ars2 in hepatocellular carcinoma.
PMID 24272391
Abstract
Hepatocellular carcinoma (HCC) is one of the most common malignant tumors in China. Arsenic resistance protein 2 (Asr2) was reported to be important for microRNA (miR) biogenesis, and its depletion could reduce the levels of several miRs, including miR-21, which is over-expressed in HCC. We hypothesized that Ars2 is also overexpressed in HCC and may be involved in the biological properties of HCC. Ars2 immunolabeling was evaluated in 132 HCCs. Ars2 immunolabeling, Ars2 qRT-PCR and miR-21 were evaluated in 20 HCCs and in paired normal tissues. Ars2 shRNA was transfected into SMCC-7721 and HepG2 HCC cells. The cell proliferation and expression of Ars2 and miR-21 were subsequently evaluated. Ars2 was expressed primarily in the nucleus of HCC cells. The expression of Ars2 was statistically correlated with the loss of HCC differentiation and pathological stage. The survival rates of patients with low Ars2 expression in HCC were statistically higher than patients with overexpressed Ars2 in HCC. Ars2 and miR-21 were more highly expressed in HCC specimens than normal tissues, and they were also correlated. The knockdown of Ars2 in HCC cells inhibited miR-21 expression and cell proliferation. Ars2 is overexpressed in HCC and may have prognostic value; it might play an important role in HCC proliferation and miR-21 expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2101297
Anti-KRAS antibody produced in rabbit, affinity isolated antibody
HPA049830
Anti-KRAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107115
Anti-KRAS antibody produced in rabbit, affinity isolated antibody
SAB4100599
Anti-KRAS antibody, Mouse monoclonal, clone KRAS-A582, purified from hybridoma cell culture
SAB2700181
Anti-NS3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501190
Anti-NSG1 antibody produced in rabbit, affinity isolated antibody
HPA042858
Anti-SRRT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503030
Anti-TCTP antibody produced in rabbit, affinity isolated antibody
R3400 Monoclonal Anti-c-K-Ras antibody produced in mouse, clone 234-4.2, purified immunoglobulin, buffered aqueous solution
WH0027065M1
Monoclonal Anti-D4S234E antibody produced in mouse, clone 1C3, purified immunoglobulin, buffered aqueous solution
WH0003845M1
Monoclonal Anti-KRAS antibody produced in mouse, clone 3B10-2F2, purified immunoglobulin, buffered aqueous solution
SAB1404011
Monoclonal Anti-KRAS antibody produced in mouse, clone 4F3, purified immunoglobulin, buffered aqueous solution
SAB4100600
Monoclonal Anti-KRAS antibody produced in mouse, clone KRAS-B582, culture supernatant