EMAIL THIS PAGE TO A FRIEND
Pediatric pulmonology
Cyclooxygenase-2 inhibition partially protects against 60% O2 -mediated lung injury in neonatal rats.
PMID 24273102
Abstract
Use of the anti-inflammatory agent dexamethasone in premature infants with bronchopulmonary dysplasia has been curtailed, and no alternative anti-inflammatory agents are approved for this use. Our objective was to use a neonatal rat model of bronchopulmonary dysplasia to determine if an highly selective cyclooxygenase-2 inhibitor, 5,5-dimethyl-3-(3-fluorophenyl)4-(4-methylsulfonyl)phenyl-2(5H)-furanone (DFU; 10 µg/g body weight), could prevent inflammatory cell influx and protect against lung injury. Neonatal rats exposed to air or 60% O2 for 14 days from birth either received daily i.p. injections of (i) vehicle or DFU or (ii) vehicle or an EP(1) receptor antagonist, SC-19220. DFU attenuated the lung macrophage and neutrophil influx, prevented interstitial thickening and prevented the loss of peripheral blood vessels induced by 60% O2 , but did not protect against the variance in alveolar diameter induced by 60% O2 . Exposure to 60% O2 caused both an increase in lung prostaglandin E2 content and a reduction in lung mesenchymal cell mass which was reversed by DFU. Prostaglandin E2 binding to the EP(1) receptor inhibited DNA synthesis in cultures of lung fibroblasts in a dose dependent fashion. Treatment with SC-19220 attenuated the reduction in lung mesenchymal mass observed following exposure of rat pups to 60% O2 . An highly selective cyclooxygenase-2 inhibitor is an effective anti-inflammatory substitute for dexamethasone for preventing phagocyte influx into the neonatal lung during 60% O2 -mediated lung injury, and can modify the severity of that injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22763 Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500247
Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405848
Anti-GAPDH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701826
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701825
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103104
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4300645
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody
HPA040067
Anti-GAPDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
D2250000
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
C20H32O5
PLA0302
Goat anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
WH0002597M1
Monoclonal Anti-GAPDH antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
P5640
Prostaglandin E2, ≥93% (HPLC), synthetic
C20H32O5
P6532
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
C20H32O5
P0409
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
C20H32O5
PLA0125
Rabbit anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.