EMAIL THIS PAGE TO A FRIEND
The Journal of pediatrics
50 years ago in the Journal of Pediatrics: the use of serum creatine phosphokinase and other serum enzymes in the diagnosis of progressive muscular dystrophy.
PMID 24274203