EMAIL THIS PAGE TO A FRIEND
Public health
Adult obesity prescribing in Wiltshire.
PMID 24275034
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O4139
Orlistat, ≥98%, solid
C29H53NO5