EMAIL THIS PAGE TO A FRIEND
Scientific reports
Is chemically synthesized graphene 'really' a unique substrate for SERS and fluorescence quenching?
PMID 24275718
Abstract
We demonstrate observation of Raman signals of different analytes adsorbed on carbonaceous materials, such as, chemically reduced graphene, graphene oxide (GO), multi-walled carbon nanotube (MWCNT), graphite and activated carbon. The analytes selected for the study were Rhodamine 6G (R6G) (in resonant conditions), Rhodamine B (RB), Nile blue (NBA), Crystal Violet (CV) and acetaminophen (paracetamol). All the analytes except paracetamol absorb and fluoresce in the visible region. In this article we provide experimental evidence of the fact that observation of Raman signals of analytes on such carbonaceous materials are more due to resonance effect, suppression of fluorescence and efficient adsorption and that this property in not unique to graphene or nanotubes but prevalent for various type of carbon materials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B2377
N-Bromoacetamide, powder
C2H4BrNO