EMAIL THIS PAGE TO A FRIEND
Organic letters
Copper-catalyzed oxidative decarboxylative couplings of sulfoximines and aryl propiolic acids.
PMID 24280013
Abstract
A method has been developed for the preparation of N-alkynylated sulfoximines involving the copper-catalyzed decarboxylative coupling of sulfoximines with aryl propiolic acids. A range of substituents on both the sulfoximidoyl moiety and the aryl group of the propiolic acid were compatible with this reaction process to afford a series of sulfoximidoyl-functionalized alkynes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR058
Continuous cast copper (O), BCR® certified Reference Material, rod
Cu
266086
Copper, powder, <425 μm, 99.5% trace metals basis
Cu
203122
Copper, powder, 99.999% trace metals basis
Cu
349178
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Cu
326453
Copper, powder (spheroidal), 10-25 μm, 98%
Cu
31284
Copper, electrolytic, Analytical Reagent, Reag. Ph. Eur., ≥99.9% (complexometric), wire
Cu
774103
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Cu
207780
Copper, powder, <75 μm, 99%
Cu
349216
Copper, wire, diam. 1.0 mm, ≥99.9%
Cu
311405
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Cu
357456
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Cu
349208
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Cu
774111
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Cu
61139
Copper, turnings, purum p.a., ≥99.0%
Cu
349151
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Cu
266744
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Cu
254177
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Cu
326488
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Cu
326429
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Cu
12816
Copper, foil, ≥99.8% (complexometric)
Cu
223409
Copper, shot, −3-+14 mesh, 99%
Cu
520381
Copper, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Cu
365327
Copper, rod, diam. 11 mm, 99.9999% trace metals basis
Cu
326410
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Cu
349224
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Cu
767476
Copper, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Cu
326437
Copper, rod, diam. 3.1 mm, 99.999%
Cu
326445
Copper, bars (random sizes), ≥99.999% trace metals basis
Cu
IRMM522C
Copper, IRMM® certified Reference Material, 0.5 mm wire
Cu
IRMM522A
Copper, IRMM® certified Reference Material, 0.1 mm foil
Cu