EMAIL THIS PAGE TO A FRIEND
Burns : Journal of the International Society For Burn Injuries
Amentoflavone inhibits angiogenesis of endothelial cells and stimulates apoptosis in hypertrophic scar fibroblasts.
PMID 24280521