EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Water-compatible hydrogen-bond activation: a scalable and organocatalytic model for the stereoselective multicomponent aza-Henry reaction.
PMID 24281805
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
803863
Schreiner′s Thiourea Catalyst, 95%
C17H8F12N2S