EMAIL THIS PAGE TO A FRIEND
Organic letters
Development of an air-stable nickel precatalyst for the amination of aryl chlorides, sulfamates, mesylates, and triflates.
PMID 24283652
Abstract
A new air-stable nickel precatalyst for C-N cross-coupling is reported. The developed catalyst system displays a greatly improved substrate scope for C-N bond formation to include both a wide range of aryl and heteroaryl electrophiles and aryl, heteroaryl, and alkylamines. The catalyst system is also compatible with a weak base, allowing the amination of substrates containing base-sensitive functional groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
802948
(dppf)Ni(o-tolyl)Cl
C41H35ClFeNiP2