EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Lower expression of the TWIK-related acid-sensitive K+ channel 2 (TASK-2) gene is a hallmark of aldosterone-producing adenoma causing human primary aldosteronism.
PMID 24285684
Abstract
The molecular mechanisms of primary aldosteronism, a common cause of human hypertension, are unknown, but alterations of K(+) channels can play a key role. The objective of the study was to investigate the following: 1) the expression of the Twik-related acid-sensitive K(+) channels (TASK) in aldosterone producing adenomas (APAs); 2) the role of TASK-2 in aldosterone synthesis; and 3) the determinants of TASK-2-blunted expression in APAs. We analyzed the transcriptome and the microRNA profiles of 32 consecutive APAs and investigated the protein expression and localization of TASK-2 in APA and adrenocortical cell lines (H295R and HAC15) using immunoblotting and confocal microscopy. The functional effect of TASK-2 blunted activity caused by a dominant-negative mutation on steroidogenic enzymes, and aldosterone production was also assessed. TASK-2 regulation by selected microRNA was studied by a luciferase assay. TASK-2 was consistently less expressed at the transcript and protein levels in APAs than in the normal human adrenal cortex. H295R cell transfection with a TASK-2 dominant-negative mutant construct significantly increased the aldosterone production by 153% and the gene expression of aldosterone synthase (CYP11B2, gene expression fold change 3.1 vs control, P < .05) and the steroidogenic acute regulatory protein (gene expression fold change 1.8 vs control, P < .05). Two microRNAs, hsa-miR-23 and hsa-miR-34, were found to decrease the TASK-2 expression by binding to the 3' untranslated region of the TASK-2 gene. The TASK-2 channel lower expression represents a hallmark of APA and is associated with a higher expression of hsa-miR-23 and hsa-miR-34. The ensuing blunted TASK-2 activity increased the production of aldosterone in vitro and the expression of steroidogenic acute regulatory protein and CYP11B2. Hence, the lower expression of TASK-2 channel in APA cells can explain high aldosterone secretion in human primary aldosteronism despite the suppression of angiotensin II, hypertension, and hypokalemia.
Related Materials