EMAIL THIS PAGE TO A FRIEND
Journal of environmental health
2013 environmental health legislation.
PMID 24288852
Abstract
The NEHA Government Affairs program has a long and productive association with the National Conference of State Legislatures (NCSL). The organizations have worked together on any number of legislative and policy areas that directly impact the environmental health profession. One of the keys to the successes of the NEHA/NCSL collaboration has been the recognition of the fact that often some of the most significant legislation and policy initiatives related to environmental public health occur in state legislatures. The states have, in a very real sense, been the innovators in developing new programs and practices. In recognition of this fact, we have asked NCSL to provide occasional overviews of state environmental public health legislative activity, covering topics that are of the most pressing public concern. Doug Farquhar, program director for NCSI's Environmental Health Program, has worked with NCSL since 1990. Mr. Farquhar directs development, management, and research for the Environmental Health Program. These projects encompass consultation and policy analysis of state and federal policies and statutes, regulations, and programs regarding environmental and related topics for state legislatures and administrative programs. Amy Ellis is a law clerk for NCSL within the Environment, Energy, and Transportation Group. As a law clerk she has researched a wide variety of environmental health policies. She is expected to obtain her JD from the University of Colorado Law School in 2015.