EMAIL THIS PAGE TO A FRIEND
MMW Fortschritte der Medizin
[Nocturnal leg calf cramps. What are the therapy options?].
PMID 24288914
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69311
Quinine, certified reference material, TraceCERT®
C20H24N2O2