EMAIL THIS PAGE TO A FRIEND
Physical review letters
Loss of collective motion in swarming bacteria undergoing stress.
PMID 24289709
Abstract
The collective motion of Bacillusxa0subtilis in the presence of a photosensitizer is disrupted by reactive oxygen species when exposed to light of sufficient dosages and is partially recovered when light irradiation is suspended. The transition from a highly collective to a more random motion is modeled using an improved self-propelled model with alignment rule. The increment in noise level describes the enhanced uncertainty in the motion of swarming bacteria under stress as observed experimentally.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
258385
Protoporphyrin IX disodium salt, ≥90% (HPLC)
C34H32N4Na2O4