EMAIL THIS PAGE TO A FRIEND
Toxicology
The role of the glutamatergic NMDA receptor in nanosilver-evoked neurotoxicity in primary cultures of cerebellar granule cells.
PMID 24291493
Abstract
Nanoparticles are known to enter the vertebrate brain, but little is known about their neurotoxicity. The aim of this study is to investigate mechanisms of the contribution of AgNPs to neuronal cell death using primary cultures of rat cerebellar granule cells (CGCs). We tested the role of glutamatergic N-methyl-d-aspartate receptors (NMDA) in AgNP-evoked neurotoxicity using MK-801, a noncompetitive inhibitor of NMDAR. We used commercially available 0.2% PVP-coated AgNPs <100 nm in a concentration range of 2.5-75 μg/ml sonicated with fetal calf serum. After a 10 min incubation period, a dose-dependent increase in the uptake of (45)Ca(2+) into neurons was observed in the presence of 25-75 μg/mL AgNPs which was completely abolished by addition of MK-801. Using the fluorescent dye fluo3 AM we observed an increase in the intracellular calcium level by 87% compared to control. ROS production was found to increase by about 30% over control after a 30-min incubation with 75 μg/mL AgNPs. Further, we observed a significant decrease in the mitochondrial potential during a 30-min incubation with AgNPs. Administration of MK-801 was found to provide a protective effect. Our results show that excitotoxicity via activation of NMDA receptor, followed by calcium imbalance, destabilization of mitochondrial function and ROS production, indicate an important mechanism involved in neurotoxicity evoked by AgNPs in cultured neurons.