EMAIL THIS PAGE TO A FRIEND
Clinical calcium
[Clinical condition and therapy of bone diseases].
PMID 24292534
Abstract
Skeletal dysplasia is the term which represents disorders including growth and differentiation of bone, cartilage and ligament. A lot of diseases are included, and new disorders have been added. However, the therapy of most bone diseases is less well-established. Achondroplasia, hypochondroplasia, and osteogenesis imperfecta are most frequent bone diseases. There is no curative treatment for these diseases, however, supportive therapies are available ; for example, growth-hormone therapy for achondroplasia and hypochondroplasia, and bisphosphonate therapy for osteogenesis imperfecta. In addition, enzyme replacement therapy for hypophosphatasia is now on clinical trial.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2371
Pamidronate disodium salt hydrate, ≥95% (NMR), solid
C3H9NO7P2Na2 · xH2O
P8361 Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
P0114
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, buffered aqueous glycerol solution, ≥6,500 DEA units/mg protein
A2356
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥5,500 DEA units/mg protein
P7923
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
P6774
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
P7640
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
P4978 Phosphatase, Alkaline from calf intestine, buffered aqueous glycerol solution
P4252 Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
P4069 Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
P5931 Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
P3895 Phosphatase, Alkaline from human placenta, ≥10 units/mg solid (in glycine buffer)
P4439 Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
A2237 Phosphatase, Alkaline shrimp, ≥1,000 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host