EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Selection of purine nucleoside analogs based on multiple biological and biochemical parameters.
PMID 2429506
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852481
6-Chloropurine riboside, 99%
C10H11ClN4O4