EMAIL THIS PAGE TO A FRIEND
Journal of helminthology
Immunogenic activity of the fish tapeworm Pterobothrium heteracanthum (Trypanorhyncha: Pterobothriidae) in BALB/c mice.
PMID 24299909
Abstract
The aim of this study was to verify the immunogenicity of Pterobothrium heteracanthum (Cestoda: Trypanorhyncha) crude protein extract (PH-CPE) in BALB/c mice. The parasites were obtained from Micropogonias furnieri (Osteichthyes: Sciaenidae). Groups of six mice were each immunized with 10, 50 or 100 μg of PH-CPE, on days 0 and 35. Both specific IgG and IgE responses were developed after immunization. The immunoblot assay revealed that specific IgG recognizes PH-CPE proteins with two molecular weight ranges, 60-75 and 30-40 kDa, and that IgE recognizes larger proteins over 120 kDa. This appears to be the first report on the immunogenicity of metacestodes within the Pterobothriidae and that PH-CPE is a potential inducer of a specific IgE response.