EMAIL THIS PAGE TO A FRIEND
Physiological Reports
Insulin signaling in skeletal muscle of HIV-infected patients in response to endurance and strength training.
PMID 24303139
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-741
Anti-AS160 (Rab-GAP) Antibody, serum, clone, Upstate®