EMAIL THIS PAGE TO A FRIEND
Aging Cell
The GATA transcription factor/MTA-1 homolog egr-1 promotes longevity and stress resistance in Caenorhabditis elegans.
PMID 24304470
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-352
Anti-acetyl-Histone H3 (Lys9) Antibody, serum, Upstate®