EMAIL THIS PAGE TO A FRIEND
Annals of hematology
Long-term follow up of frontline therapy with fludarabine and cyclophosphamide in chronic lymphocytic leukemia: impact of biological parameters on clinical outcome.
PMID 24306088