EMAIL THIS PAGE TO A FRIEND
Testing
Cognitive impairments accompanying rodent mild traumatic brain injury involve p53-dependent neuronal cell death and are ameliorated by the tetrahydrobenzothiazole PFT-?.
PMID 24312187