EMAIL THIS PAGE TO A FRIEND
Advanced pharmaceutical bulletin
Protective Role of GnRH Antagonist on Chemotherapy-induced Spermatogenesis Disorder: A Morphological Study.
PMID 24312855
Abstract
Anti cancer drugs is one of the most important chemotherapeutic factors which can influence spermatogenesis process and germinal epithelium. Since dividing cells are mainly affected by anticancer drugs, the aim of the present study is to investigate the preventive effect of GnRH antagonist on spermatogenic defect produced by anticancer drugs. In the present study thirty adult male mice aging 6-8 weeks were divided into 3 groups as: Control, Experimental 1 and Experimental 2. Experimental 1 group received Cisplatin for 5 days as 2.5 mg/kg intraperitoneally and Experimental 2 group received 0.25 mg/kg cetrorelix (GnRH antagonist) one week before cisplatin treatment and continued for 3 weeks. The mice in all groups were sacrificed 35 days after the last injection and testis specimens were fixed in boueins, formaldehyde fixative and 2.5% Glutaraldehide then prepared for light and electron microscopic examination. Light microscopy (LM) study showed that the number of spermatogonial cells, thickness of germinal epithelium, was decreased in Experimental 1group. Electron microscopy revealed that in this group several intercellular spaces appeared between spermatogenic cells and secretory granules in interstitial cells was increased. There were several vacuolated mitochondria and destroyed organelles in spermatogonial cells but in Experimental 2 group condition was similar to control group. These results indicate that the cetrorelix administration before cancer treatment may protect germinal epithelium against side effects of cisplatin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5249
Cetrorelix acetate, ≥98% (HPLC)
C70H92ClN17O14 · xC2H4O2