EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of immunology
Peripheral and site-specific CD4(+) CD28(null) T cells from rheumatoid arthritis patients show distinct characteristics.
PMID 24313359
Abstract
Proinflammatory CD4(+) CD28(null) T cells are frequently found in the circulation of patients with rheumatoid arthritis (RA), but are less common in the rheumatic joint. In the present study, we sought to identify functional differences between CD4(+) CD28(null) T cells from blood and synovial fluid in comparison with conventional CD28-expressing CD4(+) T cells. Forty-four patients with RA, displaying a distinct CD4(+) CD28(null) T cell population in blood, were recruited for this study; the methylation status of the IFNG locus was examined in isolated T cell subsets, and intracellular cytokine production (IFN-γ, TNF, IL-17) and chemokine receptor expression (CXCR3, CCR6 and CCR7) were assessed by flow cytometry on T cells from the two compartments. Circulating CD4(+) CD28(null) T cells were significantly more hypomethylated in the CNS-1 region of the IFNG locus than conventional CD4(+) CD28(+) T cells and produced higher levels of both IFN-γ and TNF after TCR cross-linking. CD4(+) CD28(null) T cells from the site of inflammation expressed significantly more CXCR3 and CCR6 compared to their counterparts in blood. While IL-17A production could hardly be detected in CD4(+) CD28(null) cells from the blood, a significant production was observed in CD4(+) CD28(null) T cells from synovial fluid. CD4(+) CD28(null) T cells were not only found to differ from conventional CD4(+) CD28(+) T cells in the circulation, but we could also demonstrate that synovial CD4(+) CD28(null) T cells showed additional effector functions (IL-17 coproduction) as compared to the same subset in peripheral blood, suggesting an active role for these cells in the perpetuation of inflammation in the subset of patients having a CD28(null) population.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3241 Anti-CCR-7 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4500329
Anti-CCR7 antibody produced in rabbit, affinity isolated antibody
SAB4300627
Anti-CCR7 antibody produced in rabbit, affinity isolated antibody
GW22783 Anti-CD14 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500215
Anti-CD14 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405579
Anti-CD14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001887
Anti-CD14 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002127
Anti-CD14 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501381
Anti-Cd28 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501674
Anti-CD28 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405583
Anti-CD28 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
C7831 Anti-CD28 antibody, Mouse monoclonal, clone CD28.2, purified from hybridoma cell culture
HPA004252
Anti-CD4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004472
Anti-CD4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
F1773 Anti-CD4−FITC antibody, Mouse monoclonal, clone Q4120, purified from hybridoma cell culture
SAB4700103 Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-18, purified immunoglobulin, buffered aqueous solution
SAB4700669 Monoclonal Anti-CD14 antibody produced in mouse, clone B-A8, purified immunoglobulin, buffered aqueous solution
SAB4700097
Monoclonal Anti-CD14 antibody produced in mouse, clone MEM-15, purified immunoglobulin, buffered aqueous solution
C7673
Monoclonal Anti-CD14 antibody produced in mouse, clone UCHM-1, purified immunoglobulin, buffered aqueous solution
AMAB90897
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1637, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90898
Monoclonal Anti-CD14 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1638, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700104
Monoclonal Anti-CD14-APC antibody produced in mouse, clone MEM-18, purified immunoglobulin, buffered aqueous solution
SAB4700098 Monoclonal Anti-CD14-APC antibody produced in mouse, clone MEM-15, purified immunoglobulin, buffered aqueous solution
SAB4700100
Monoclonal Anti-CD14-FITC antibody produced in mouse, clone MEM-15, purified immunoglobulin, buffered aqueous solution
SAB4700106
Monoclonal Anti-CD14-FITC antibody produced in mouse, clone MEM-18, purified immunoglobulin, buffered aqueous solution
SAB4700110
Monoclonal Anti-CD19-APC antibody produced in mouse, clone LT19, purified immunoglobulin, buffered aqueous solution
SAB4700717 Monoclonal Anti-CD19-APC antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
SAB4700112 Monoclonal Anti-CD19-FITC antibody produced in mouse, clone LT19, purified immunoglobulin, buffered aqueous solution
SAB4700704 Monoclonal Anti-CD19-FITC antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
SAB4700135 Monoclonal Anti-CD28 antibody produced in mouse, low endotoxin, clone CD28.2, purified immunoglobulin, buffered aqueous solution