EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
Leishmania amazonensis: characterization of an ecto-pyrophosphatase activity.
PMID 24316462
Abstract
Several ecto-enzymatic activities have been described in the plasma membrane of the protozoan Leishmania amazonensis, which is the major etiological agent of diffuse cutaneous leishmaniasis in South America. These enzymes, including ecto-phosphatases, contribute to the survival of the parasite by participating in phosphate metabolism. This work identifies and characterizes the extracellular hydrolysis of inorganic pyrophosphate related to an ecto-pyrophosphatase activity of the promastigote form of L. amazonensis. This ecto-pyrophosphatase activity is insensitive to MnCl2 but is strongly stimulated by MgCl2. This stimulation was not observed during the hydrolysis of p-nitrophenyl phosphate (p-NPP) or β-glycerophosphate, two substrates for different ecto-phosphatases present in the L. amazonensis plasma membrane. Furthermore, extracellular PPi hydrolysis is more efficient at alkaline pHs, while p-NPP hydrolysis occurs mainly at acidic pHs. These results led us to conclude that extracellular PPi is hydrolyzed not by non-specific ecto-phosphatases but rather by a genuine ecto-pyrophosphatase. In the presence of 5mM MgCl2, the ecto-pyrophosphatase activity from L. amazonensis is sensitive to micromolar concentrations of NaF and millimolar concentrations of CaCl2. Moreover, this activity is significantly higher during the first days of L. amazonensis culture, which suggests a possible role for this enzyme in parasite growth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L-025
(−)-Levamisole hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material
C11H12N2S
L9756
(−)-Tetramisole hydrochloride, ≥99% (GC)
C11H12N2S · HCl
223506
Calcium chloride dihydrate, ACS reagent, ≥99%
CaCl2 · 2H2O
C3881
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
CaCl2 · 2H2O
31307
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
CaCl2 · 2H2O
12022
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
CaCl2 · 2H2O
T206
D-(−)-Tartaric acid, ReagentPlus®, 99%
C4H6O6
483796
D-(−)-Tartaric acid, 99%
C4H6O6
95320
D-(−)-Tartaric acid, puriss., unnatural form, ≥99.0% (T)
C4H6O6
03918
DL-Tartaric acid, anhydrous, for ion chromatography, ≥99.5% (T)
C4H6O6
T400
DL-Tartaric acid, ReagentPlus®, 99%
C4H6O6
W304401
DL-Tartaric acid, ≥99%
C4H6O6
251380
L-(+)-Tartaric acid, ACS reagent, ≥99.5%
C4H6O6
W304409
L-(+)-Tartaric acid, ≥99.7%, FCC, FG
C4H6O6
T109
L-(+)-Tartaric acid, ≥99.5%
C4H6O6
T1807
L-(+)-Tartaric acid, BioXtra
C4H6O6
33801
L-(+)-Tartaric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)
C4H6O6
41447
L-(+)-Tartaric acid, certified reference material, TraceCERT®
C4H6O6
27506
L-(+)-Tartaric acid, puriss., meets analytical specification of Ph. Eur., NF, 99.7-100.5% (calc. to the dried substance), powder
C4H6O6
95318
L-(+)-Tartaric acid, tested according to Ph.Eur.
C4H6O6
798665
L-(+)-Tartaric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
C4H6O6
31742
Levamisol hydrochloride, VETRANAL, analytical standard
C11H12N2S · HCl
L0380000
Levamisole hydrochloride, European Pharmacopoeia (EP) Reference Standard
C11H12N2S · HCl
Y0000047
Levamisole hydrochloride for system suitability, European Pharmacopoeia (EP) Reference Standard
C11H12N2S · HCl
221279
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Cl2Mn · 4H2O
M8054
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Cl2Mn · 4H2O
203734
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Cl2Mn · 4H2O
M3634
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Cl2Mn · 4H2O
M5005
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Cl2Mn · 4H2O
63535
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Cl2Mn · 4H2O