EMAIL THIS PAGE TO A FRIEND
FEBS letters
Downregulation of histone deacetylase 1 by microRNA-520h contributes to the chemotherapeutic effect of doxorubicin.
PMID 24316511
Abstract
Doxorubicin induces DNA damage to exert its anti-cancer function. Histone deacetylase 1 (HDAC1) can protect the genome from DNA damage. We found that doxorubicin specifically downregulates HDAC1 protein expression and identified HDAC1 as a target of miR-520h, which was upregulated by doxorubicin. Doxorubicin-induced cell death was impaired by exogenous HDAC1 or by miR-520h inhibitor. Moreover, HDAC1 reduced the level of γH2AX by preventing the interaction of doxorubicin with DNA. In summary, doxorubicin downregulates HDAC1 protein expression, by inducing the expression of HDAC1-targeting miR-520h, to exacerbate DNA-doxorubicin interaction. The upregulation of HDAC1 protein may contribute to drug resistance of human cancer cells and targeting HDAC1 is a promising strategy to increase the clinical efficacy of DNA damage-inducing chemotherapeutic drugs.