EMAIL THIS PAGE TO A FRIEND
Anticancer research
Comparison of minichromosome maintenance proteins (MCM-3, MCM-7) and metallothioneins (MT-I/II, MT-III) expression in relation to clinicopathological data in ovarian cancer.
PMID 24324072
Abstract
Despite great progress in the understanding of ovarian cancer biology, clinicopathological data (i.e. grade, stage, histological type and residual disease after surgery) seem to be the most important prognostic factors. The present study aimed to investigate the relationship between expression of minichromosome maintenance proteins (MCM-3, MCM-7), metallothioneins (MT-I/II, MT-III), and Ki-67 in 103 ovarian cancer cases, mostly of the serous histological type. Statistical analysis revealed strong positive correlations in the expression of MCM-3 vs. Ki-67 (r=0.492), MCM-7 vs. Ki-67 (r=0.651), and MCM-3 vs. MCM-7 (r=0.515) (all p<0.0001). The Kruskal-Wallis test showed an association of increased expression of MCM-3 and Ki-67 with increasing grade of histological malignancy (p=0.0011, p=0.029, respectively). Regarding clinical progression, cytoplasmic MT-I/II expression was significantly higher in more advanced disease stages (III+IV vs. I+II; p=0.0247). Due to the correlations shown here, the determination of MCM proteins as proliferation markers of ovarian cancer, should be strongly considered.