EMAIL THIS PAGE TO A FRIEND
Journal of tropical pediatrics
Acute disseminated encephalomyelitis presenting with hypertensive emergency.
PMID 24327453
Abstract
We report a 12-year-old girl presenting with acute disseminated encephalomyelitis (ADEM) along with hypertensive emergency. Hypertension persisted for few weeks following recovery and subsided with oral clonidine. Although autonomic instability in ADEM has been reported before, hypertensive emergency was not previously documented as presenting feature of ADEM.
Related Materials