EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Neurosteroid analogues. 18. Structure-activity studies of ent-steroid potentiators of γ-aminobutyric acid type A receptors and comparison of their activities with those of alphaxalone and allopregnanolone.
PMID 24328079
Abstract
A model of the alignment of neurosteroids and ent-neurosteroids at the same binding site on γ-aminobutyric acid type A (GABAA) receptors was evaluated for its ability to identify the structural features in ent-neurosteroids that enhance their activity as positive allosteric modulators of this receptor. Structural features that were identified included: (1) a ketone group at position C-16, (2) an axial 4α-OMe group, and (3) a C-18 methyl group. Two ent-steroids were identified that were more potent than the anesthetic steroid alphaxalone in their threshold for and duration of loss of the righting reflex in mice. In tadpoles, loss of righting reflex for these two ent-steroids occurs with EC50 values similar to those found for allopregnanolone. The results indicate that ent-steroids have considerable potential to be developed as anesthetic agents and as drugs to treat brain disorders that are ameliorated by positive allosteric modulators of GABAA receptor function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8129
5β-Pregnan-3α-ol-20-one
C21H34O2