EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Release and metabolism of dopamine and serotonin in the caudate nucleus during euthermia and hibernation.
PMID 2432821
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
148830
Homovanillyl alcohol, 99%
C9H12O3