EMAIL THIS PAGE TO A FRIEND
Clinical immunology (Orlando, Fla.)
Protective role of JAK/STAT signaling against renal fibrosis in mice with unilateral ureteral obstruction.
PMID 24333535
Abstract
Inflammation is involved in renal fibrosis, a final common pathway for kidney diseases. To clarify how JAK/STAT/SOCS system was involved in renal fibrosis, UUO was induced in BALB/c or SOCS3(+/-) mice in the presence or absence of JAK inhibitor-incorporated nanoparticle (pyridine6-PGLA). UUO increased pSTAT3 and subsequently elevated SOCS3 levels in the obstructed kidneys. pSTAT3 levels were further increased in SOCS3(+/-) mice. UUO-induced renal fibrosis was markedly suppressed in SOCS3(+/-) mice, while it was aggravated by pre-treatment with pyridine6-PGLA. Although there were no differences in renal mRNA levels of TGF-β and collagens between wild and SOCS3(+/-) mice, MMP-2 activity was enhanced in SOCS3(+/-) UUO mice. Activated MMP-2 was completely suppressed by pyridine6-PGLA-pre-treatment. TNF-α one of JAK/STAT activators, increased pSTAT3 levels and subsequently induced MMP-2 activation in proximal tubular cells. These results suggest that JAK/STAT3 signaling may play a role in repair process of renal fibrosis in UUO partly via MMP-2 activation.