EMAIL THIS PAGE TO A FRIEND
HIV clinical trials
Addition of nitazoxanide to PEG-IFN and ribavirin to improve HCV treatment response in HIV-1 and HCV genotype 1 coinfected persons naïve to HCV therapy: results of the ACTG A5269 trial.
PMID 24334180
Abstract
We hypothesized that nitazoxanide (NTZ) added to pegylated interferon alfa-2a (PEG-IFN) and weight-based ribavirin (WBR) would improve hepatitis C virus (HCV) virologic responses in HCV treatment-naïve HIV-1/HCV genotype 1 coinfected persons. Prospective, single-arm study in which subjects received 4-week lead-in (NTZ 500 mg twice daily) followed by 48 weeks of NTZ, PEG-IFN, and WBR. We compared the HCV virologic responses of these subjects to historical controls from the completed ACTG study A5178 who received PEG-IFN and WBR and had similar subject characteristics. Primary endpoints were early virologic response and complete early virologic response (EVR and cEVR). Among 67 subjects (78% male; 48% Black; median age, 50 years), EVR was achieved in 65.7% (90% CI, 55.0%-75.3%), cEVR in 38.8% (28.8%-49.6%). and SVR in 32.8% (23.4%-43.5%). EVR was higher with NTZ (51.4% in A5178; P = .03), but the sustained virologic response (SVR) proportion was similar (27.3% in A5178; P = .24). In contrast to A5178, SVR was similar across IL28B genotypes. Overall, NTZ was safe and well-tolerated. Whereas EVR proportion improved significantly in this pilot study, the addition of NTZ to PEG-IFN/WBR did not significantly improve SVR compared to historical controls. NTZ may be associated with an attenuation of the effect of IL28B on HCV treatment response.