EMAIL THIS PAGE TO A FRIEND
Recenti progressi in medicina
[Incomplete atrioventricular block in a patient on pregabalin therapy].
PMID 24336619
Abstract
The authors report on a case of incomplete atrio-ventricular block in a patient on pregabalin therapy. Pregabalin was not overdosed; renal function of the patient was normal. The effect reverted after pregabalin discontinuation.